APO455832593 Apollo Scientific Ltd

N-(1,4-Benzodioxan-2-carbonyl)piperazine hydrochloride

  • MDL number MFCD00729051
  • Cas Number : 70918-74-0
  • Molecular Weight : 284.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 263
Purity 98
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/n-14-benzodioxan-2-carbonylpiperazine-hydrochloride

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/n-14-benzodioxan-2-carbonylpiperazine-hydrochloride
pictograms GHS06;GHS08;GHS09
hstatements H301+H311+H331;H373
Availability details for
Quantity
  • Availability
  • Add to Cart