APO455832594 Apollo Scientific Ltd

3-Bromo-7-nitroindazole

  • MDL number MFCD00159910
  • Cas Number : 74209-34-0
  • Molecular Weight : 242.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 177-178
Iupac Name 3-bromo-7-nitro-1H-indazole
Inchi Code 1S/C7H4BrN3O2/c8-7-4-2-1-3-5(11(12)13)6(4)9-10-7/h1-3H,(H,9,10)
Inchi Key NFSTZPMYAZRZPC-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 177-178
Product Link https://store.apolloscientific.co.uk/product/3-bromo-7-nitroindazole

Safety Information

signalword Danger
pcodes P280;P301+P312;P308+P313
MSDS https://store.apolloscientific.co.uk/product/3-bromo-7-nitroindazole
pictograms GHS07;GHS08
hstatements H302;H360
Availability details for
Quantity
  • Availability
  • Add to Cart