APO455832607 Apollo Scientific Ltd

4-[(4,6-Dimethylpyrimidin-2-yl)(methyl)amino]benzoic acid

  • MDL number MFCD00203268
  • Cas Number : 387350-50-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 172-174
Synonym 4-[(4,6-Dimethyl-1,3-diazin-2-yl)(methyl)amino]benzoic acid, 2-[(4-Carboxyphenyl)(methyl)amino]-4,6-dimethylpyrimidine
Purity 95
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=14496

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=14496
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart