APO455832615 Apollo Scientific Ltd

1-(Cyclohexylmethyl)piperazine

  • MDL number MFCD01075237
  • Cas Number : 57184-23-3
  • Molecular Weight : 182.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Density 0.93
Flash Point 87.8
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 230-231
Product Link https://store.apolloscientific.co.uk/product/1-cyclohexylmethylpiperazine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1-cyclohexylmethylpiperazine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart