APO455832616 Apollo Scientific Ltd

1-(Cycloheptyl)piperazine

  • MDL number MFCD01076573
  • Cas Number : 21043-42-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 102/2.5mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=14019

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=14019
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart