APO455832618 Apollo Scientific Ltd

1-(2-Diallylaminoethyl)piperazine

  • MDL number MFCD00191216
  • Cas Number : 199475-35-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N-[2-(Piperazin-1-yl)ethyl]-N-(prop-2-en-1-yl)prop-2-en-1-amine
Purity 97
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 76-78/0.5mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=14087

Safety Information

signalword Danger
pcodes P260;P280;P309+P311
MSDS http://www.apolloscientific.co.uk/display_item.php?id=14087
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart