APO455832631 Apollo Scientific Ltd

1-Methylpiperidine-4-carboxylic acid hydrochloride

  • MDL number MFCD00190235
  • Cas Number : 71985-80-3
  • Molecular Weight : 179.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 225
Iupac Name 1-methyl-4-piperidinecarboxylic acid hydrochloride
Inchi Code 1S/C7H13NO2.ClH/c1-8-4-2-6(3-5-8)7(9)10;/h6H,2-5H2,1H3,(H,9,10);1H
Inchi Key NLUDEWJJEMHIIL-UHFFFAOYSA-N

Description

Synonym 4-Carboxy-1-methylpiperidine hydrochloride, 1-Methylisonipecotic acid hydrochloride
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 225
Product Link https://store.apolloscientific.co.uk/product/1-methylpiperidine-4-carboxylic-acid-hydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-methylpiperidine-4-carboxylic-acid-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart