APO455832637 Apollo Scientific Ltd

Methyl 8-hydroxy-1-naphthoate

  • MDL number MFCD00665832
  • Cas Number : 5247-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Methyl 8-hydroxynaphthalene-1-carboxylate
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=15217

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=15217
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart