APO455832649 Apollo Scientific Ltd

1-Bromo-2,4-dimethyl-5-nitrobenzene

  • MDL number MFCD00075824
  • Cas Number : 69383-59-1
  • Molecular Weight : 230.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 55-57
Iupac Name 1-bromo-2,4-dimethyl-5-nitrobenzene
Inchi Code 1S/C8H8BrNO2/c1-5-3-6(2)8(10(11)12)4-7(5)9/h3-4H,1-2H3
Inchi Key HYYJANCKMGXHLQ-UHFFFAOYSA-N

Description

Synonym 4-Bromo-6-nitro-m-xylene, 5-Bromo-2,4-dimethylnitrobenzene
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 55-57
Product Link https://store.apolloscientific.co.uk/product/1-bromo-24-dimethyl-5-nitrobenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1-bromo-24-dimethyl-5-nitrobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart