APO455832650 Apollo Scientific Ltd

2,4-Dimethyl-5-nitroaniline

  • MDL number MFCD00025199
  • Cas Number : 2124-47-2
  • Molecular Weight : 166.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Amino-2,4-dimethylnitrobenzene, 5-Nitro-2,4-xylidine
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/24-dimethyl-5-nitroaniline

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/24-dimethyl-5-nitroaniline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart