APO455832653 Apollo Scientific Ltd

4-Acetamido-3-ethylbenzonitrile

  • MDL number MFCD00221461
  • Cas Number : 34921-76-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178-180
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=12341

Safety Information

signalword Warning
pcodes P280;P271;P261
MSDS http://www.apolloscientific.co.uk/display_item.php?id=12341
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart