APO455832658 Apollo Scientific Ltd

2'-Sulphamoyl-[1,1'-biphenyl]-4-carboxylic acid

  • MDL number MFCD03001764
  • Cas Number : 352615-90-8
  • Molecular Weight : 277.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2'-(aminosulfonyl)[1,1'-biphenyl]-4-carboxylic acid
Inchi Code 1S/C13H11NO4S/c14-19(17,18)12-4-2-1-3-11(12)9-5-7-10(8-6-9)13(15)16/h1-8H,(H,15,16)(H2,14,17,18)
Inchi Key LYHKPAIFSGHBPU-UHFFFAOYSA-N

Description

Synonym 2'-(Aminosulphonyl)-[1,1'-biphenyl]-4-carboxylic acid, 4-(2-Sulphamoylphenyl)benzoic acid
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-sulphamoyl-11-biphenyl-4-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-sulphamoyl-11-biphenyl-4-carboxylic-acid
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart