APO455832666 Apollo Scientific Ltd

4-Bromo-2-(methylsulphonyl)aniline

  • MDL number MFCD21193892
  • Cas Number : 1396554-49-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130-132
Synonym 2-Amino-5-bromophenyl methyl sulphone
Storage temp. Ambient temperature
Physical Form Solid-Crystals
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=51003

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=51003
pictograms GHS07
hstatements H302+H312+H332;H335
Availability details for
Quantity
  • Availability
  • Add to Cart