APO455832673 Apollo Scientific Ltd

2-Bromo-3-methoxyaniline

  • MDL number MFCD07787450
  • Cas Number : 112970-44-2
  • Molecular Weight : 202.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Bromo-m-anisidine, 3-Amino-2-bromoanisole
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-3-methoxyaniline

Safety Information

signalword Danger
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-3-methoxyaniline
pictograms GHS06;GHS07
hstatements H301+H311+H331;H315;H319
Availability details for
Quantity
  • Availability
  • Add to Cart