APO455832674 Apollo Scientific Ltd

3-(Hydroxymethyl)tetrahydro-2H-pyran

  • MDL number MFCD08235011
  • Cas Number : 14774-36-8
  • Molecular Weight : 116.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tetrahydro-2H-pyran-3-ylmethanol
density 1.0553
Inchi Code 1S/C6H12O2/c7-4-6-2-1-3-8-5-6/h6-7H,1-5H2
Inchi Key VFTQJKSRDCKVQA-UHFFFAOYSA-N

Description

Synonym (Tetrahydro-2H-pyran-3-yl)methanol, 3-(Hydroxymethyl)oxane
Density 1.0553
Flash Point 81/4mm
Storage temp. Store at 2-8
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-hydroxymethyltetrahydro-2h-pyran

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-hydroxymethyltetrahydro-2h-pyran
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart