APO455832676 Apollo Scientific Ltd

Tetrahydro-2H-pyran-3-carboxaldehyde

  • MDL number MFCD03789633
  • Cas Number : 77342-93-9
  • Molecular Weight : 114.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Formyltetrahydro-2H-pyran, 3-Formyloxane
Density 1.06
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 54-55/7mm
Product Link https://store.apolloscientific.co.uk/product/tetrahydro-2h-pyran-3-carboxaldehyde

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/tetrahydro-2h-pyran-3-carboxaldehyde
pictograms GHS05;GHS07
hstatements H302;H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart