APO455832679 Apollo Scientific Ltd

1-(4-Bromophenyl)naphthalene

  • MDL number MFCD07369916
  • Cas Number : 204530-94-9
  • Molecular Weight : 283.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-4-bromophenylnaphthalene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-4-bromophenylnaphthalene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart