APO455832680 Apollo Scientific Ltd

4-Bromo-4'-methylbiphenyl

  • MDL number MFCD00466236
  • Cas Number : 50670-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 136
Synonym 4-(4-Bromophenyl)toluene
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=61447

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=61447
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart