APO455832686 Apollo Scientific Ltd

5-Chloro-2-iodoanisole

  • MDL number MFCD09753735
  • Cas Number : 755027-21-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 20
Synonym 4-Chloro-1-iodo-2-methoxybenzene, 5-Chloro-2-iodophenyl methyl ether
Purity 97
Storage temp. Ambient temperature
Physical Form Liquid-Clear
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 135/10mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=61456

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=61456
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart