APO455832690 Apollo Scientific Ltd

2-Bromo-3-chlorobenzoic acid

  • MDL number MFCD00672931
  • Cas Number : 56961-26-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143-146
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=61624

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=61624
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart