APO455832701 Apollo Scientific Ltd

2-Bromo-3-iodoanisole

  • MDL number MFCD18391616
  • Cas Number : 74128-84-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 43-47
Synonym 2-Bromo-1-iodo-3-methoxybenzene, 2-Bromo-3-iodophenyl methyl ether
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=61500

Safety Information

signalword Warning
msds http://www.apolloscientific.co.uk/display_item.php?id=61500
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart