APO455832703 Apollo Scientific Ltd

4-Isopropylbenzene-1,3-diol

  • MDL number MFCD05663778
  • Cas Number : 23504-03-2
  • Molecular Weight : 152.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Isopropylresorcinol, 1,3-Dihydroxy-4-(prop-2-yl)benzene
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-isopropylbenzene-13-diol

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-isopropylbenzene-13-diol
pictograms GHS05;GHS07
hstatements H302;H317;H314
Availability details for
Quantity
  • Availability
  • Add to Cart