APO455832711 Apollo Scientific Ltd

2-Bromo-4-chloro-6-nitroaniline

  • MDL number MFCD08275709
  • Cas Number : 827-25-8
  • Molecular Weight : 251.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-4-chloro-6-nitroaniline
Inchi Code 1S/C6H4BrClN2O2/c7-4-1-3(8)2-5(6(4)9)10(11)12/h1-2H,9H2
Inchi Key NPCIWMOFULRTDX-UHFFFAOYSA-N

Description

Synonym 2-Amino-3-bromo-5-chloronitrobenzene
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-4-chloro-6-nitroaniline

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-4-chloro-6-nitroaniline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart