APO455832722 Apollo Scientific Ltd

Bicyclo[4.1.0]heptane-7-carboxylic acid

  • MDL number MFCD00095180
  • Cas Number : 41894-76-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 7-Carboxybicyclo[4.1.0]heptane, Norcarane-7-carboxylic acid
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=26843

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=26843
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart