APO455832723 Apollo Scientific Ltd

4-Bromo-5-methylisatin

  • MDL number MFCD00560709
  • Cas Number : 147149-84-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Bromo-5-methyl-1H-indole-2,3-dione, 4-Bromo-5-methylindolin-2,3-dione
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-bromo-5-methylisatin

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-5-methylisatin
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart