APO455832729 Apollo Scientific Ltd

4-(2-Bromoethyl)-3,5-dimethyl-1H-pyrazole

  • MDL number MFCD00480987
  • Cas Number : 83467-28-1
  • Molecular Weight : 203.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 118-120
Iupac Name 4-(2-bromoethyl)-3,5-dimethyl-1H-pyrazole
Inchi Code 1S/C7H11BrN2/c1-5-7(3-4-8)6(2)10-9-5/h3-4H2,1-2H3,(H,9,10)
Inchi Key HGGLMDBNLJUEKA-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 118-120
Product Link https://store.apolloscientific.co.uk/product/4-2-bromoethyl-35-dimethyl-1h-pyrazole

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-2-bromoethyl-35-dimethyl-1h-pyrazole
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart