APO455832734 Apollo Scientific Ltd

4,4'-Bipyridine dihydrochloride

  • MDL number MFCD00012824
  • Cas Number : 27926-72-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 302-306
Density 1.52
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=26886

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=26886
pictograms GHS06
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart