APO455832735 Apollo Scientific Ltd

5-Formylthiophene-2-carboxylic acid

  • MDL number MFCD00191437
  • Cas Number : 4565-31-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169(dec.)
Synonym 5-Carboxythiophene-2-carboxaldehyde, 2-Carboxy-5-formylthiophene, 5-Formyl-2-thenoic acid
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-formylthiophene-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-formylthiophene-2-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart