APO455832736 Apollo Scientific Ltd

3-Methylisoxazole-4-carboxylic acid

  • MDL number MFCD01318161
  • Cas Number : 17153-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 183-190
Synonym 3-Methyl-1,2-oxazole-4-carboxylic acid, 4-Carboxy-3-methylisoxazole
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-methylisoxazole-4-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-methylisoxazole-4-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart