APO455832737 Apollo Scientific Ltd

3-Phenylcyclobutan-1-one

  • MDL number MFCD04038993
  • Cas Number : 52784-31-3
  • Molecular Weight : 146.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-phenylcyclobutanone
Inchi Code 1S/C10H10O/c11-10-6-9(7-10)8-4-2-1-3-5-8/h1-5,9H,6-7H2
Inchi Key BVQSFCUGCAZOJQ-UHFFFAOYSA-N

Description

Synonym 1-Oxo-3-phenylcyclobutane
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-phenylcyclobutan-1-one

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-phenylcyclobutan-1-one
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart