APO455832738 Apollo Scientific Ltd

5-Chloronaphthalene-1-sulphonyl chloride

  • MDL number MFCD00269949
  • Cas Number : 6291-07-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94-95
Synonym 5-Chloro-1-(chlorosulphonyl)naphthalene
Storage temp. Store at -20
Physical Form Solid
Shipping Temperature Shipped with Ice Packs
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-chloronaphthalene-1-sulphonyl-chloride

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-chloronaphthalene-1-sulphonyl-chloride
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart