APO455832739 Apollo Scientific Ltd

(1H-1,2,4-Triazol-1-yl)acetic acid

  • MDL number MFCD00036051
  • Cas Number : 28711-29-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 201-205
Synonym 2-(1H-1,2,4-Triazol-1-yl)ethanoic acid
Purity 98
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=26898

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=26898
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart