APO455832740 Apollo Scientific Ltd

1-(2-Aminoethyl)-1H-1,2,4-triazole dihydrochloride

  • MDL number MFCD08445540
  • Cas Number : 51444-26-9
  • Molecular Weight : 185.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(1H-1,2,4-triazol-1-yl)ethanamine dihydrochloride
Inchi Code 1S/C4H8N4.2ClH/c5-1-2-8-4-6-3-7-8;;/h3-4H,1-2,5H2;2*1H
Inchi Key DTPGFYKFVNFLLK-UHFFFAOYSA-N

Description

Synonym 2-(1H-1,2,4-Triazol-1-yl)ethylamine dihydrochloride
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-2-aminoethyl-1h-124-triazole-dihydrochloride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-2-aminoethyl-1h-124-triazole-dihydrochloride
pictograms GHS07
hstatements H302+H312+H332;H319
Availability details for
Quantity
  • Availability
  • Add to Cart