APO455832741 Apollo Scientific Ltd

1-(3-Aminoprop-1-yl)-1H-1,2,4-triazole hydrochloride

  • MDL number MFCD06796387
  • Cas Number : 794522-91-1
  • Molecular Weight : 162.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(1H-1,2,4-Triazol-1-yl)propylamine hydrochloride, 1-Amino-3-(1H-1,2,4-triazol-1-yl)propane hydrochloride
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-3-aminoprop-1-yl-1h-124-triazole-hydrochloride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1-3-aminoprop-1-yl-1h-124-triazole-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart