APO455832756 Apollo Scientific Ltd

Benzo[b]furan-5-boronic acid

  • MDL number MFCD04038999
  • Cas Number : 331834-13-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 216-220
Synonym 1-Benzofuran-5-boronic acid, 5-Borono-1-benzofuran
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/benzobfuran-5-boronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/benzobfuran-5-boronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart