APO455832761 Apollo Scientific Ltd

2-Bromo-5-pyridylboronic acid pinacol ester

  • MDL number MFCD03412794
  • Cas Number : 214360-62-0
  • Molecular Weight : 283.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Inchi Code 1S/C11H15BBrNO2/c1-10(2)11(3,4)16-12(15-10)8-5-6-9(13)14-7-8/h5-7H,1-4H3
Inchi Key MEGBELYYUPCOIQ-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-5-pyridylboronic-acid-pinacol-ester

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-5-pyridylboronic-acid-pinacol-ester
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart