APO455832764 Apollo Scientific Ltd

3-(tert-Butoxymethyl)benzeneboronic acid

  • MDL number MFCD03788411
  • Cas Number : 858364-78-0
  • Molecular Weight : 208.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(tert-butoxymethyl)phenylboronic acid
Inchi Code 1S/C11H17BO3/c1-11(2,3)15-8-9-5-4-6-10(7-9)12(13)14/h4-7,13-14H,8H2,1-3H3
Inchi Key XAAQNVFGBHTNRX-UHFFFAOYSA-N

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-tert-butoxymethylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-tert-butoxymethylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart