APO455832771 Apollo Scientific Ltd

3-Chloropyridine-4-boronic acid

  • MDL number MFCD03425942
  • Cas Number : 458532-98-4
  • Molecular Weight : 157.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 95-97
Iupac Name 3-chloro-4-pyridinylboronic acid
Inchi Code 1S/C5H5BClNO2/c7-5-3-8-2-1-4(5)6(9)10/h1-3,9-10H
Inchi Key JLIHQPWLAOYTRH-UHFFFAOYSA-N

Description

Synonym 4-Borono-3-chloropyridine
Storage temp. Store at -20
Physical Form Solid
Shipping Temperature Shipped with Ice Packs
Country of Origin GB
Boiling point 95-97
Product Link https://store.apolloscientific.co.uk/product/3-chloropyridine-4-boronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloropyridine-4-boronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart