APO455832778 Apollo Scientific Ltd

3,5-Dichloro-4-methoxybenzeneboronic acid

  • MDL number MFCD04039018
  • Cas Number : 175883-61-1
  • Molecular Weight : 220.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3,5-dichloro-4-methoxyphenylboronic acid
Inchi Code 1S/C7H7BCl2O3/c1-13-7-5(9)2-4(8(11)12)3-6(7)10/h2-3,11-12H,1H3
Inchi Key JHCLSOGJYUVJNQ-UHFFFAOYSA-N

Description

Synonym 4-Borono-2,6-dichloroanisole
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/35-dichloro-4-methoxybenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/35-dichloro-4-methoxybenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart