APO455832785 Apollo Scientific Ltd

4-(Heptyloxy)benzeneboronic acid

  • MDL number MFCD04039169
  • Cas Number : 136370-19-9
  • Molecular Weight : 236.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(heptyloxy)phenylboronic acid
Inchi Code 1S/C13H21BO3/c1-2-3-4-5-6-11-17-13-9-7-12(8-10-13)14(15)16/h7-10,15-16H,2-6,11H2,1H3
Inchi Key UHRONCCLURKEKO-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-heptyloxybenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-heptyloxybenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart