APO455832789 Apollo Scientific Ltd

4-[(Tetrahydro-2H-pyran-2-yl)oxy]benzeneboronic acid

  • MDL number MFCD02183525
  • Cas Number : 182281-01-2
  • Molecular Weight : 222.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 116-123
Iupac Name 4-(tetrahydro-2H-pyran-2-yloxy)phenylboronic acid
Inchi Code 1S/C11H15BO4/c13-12(14)9-4-6-10(7-5-9)16-11-3-1-2-8-15-11/h4-7,11,13-14H,1-3,8H2
Inchi Key DMGMCZRNMYQQQB-UHFFFAOYSA-N

Description

Synonym 2-(4-Boronophenoxy)tetrahydro-2H-pyran
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 116-123
Product Link https://store.apolloscientific.co.uk/product/4-tetrahydro-2h-pyran-2-yloxybenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-tetrahydro-2h-pyran-2-yloxybenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart