APO455832795 Apollo Scientific Ltd

3-(Methoxymethyl)benzeneboronic acid

  • MDL number MFCD03412069
  • Cas Number : 142273-84-5
  • Molecular Weight : 165.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(methoxymethyl)phenylboronic acid
Inchi Code 1S/C8H11BO3/c1-12-6-7-3-2-4-8(5-7)9(10)11/h2-5,10-11H,6H2,1H3
Inchi Key XOSGESMDNPVZKS-UHFFFAOYSA-N

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-methoxymethylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-methoxymethylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart