APO455832804 Apollo Scientific Ltd

4-(n-Pentyloxy)benzeneboronic acid

  • MDL number MFCD04039035
  • Cas Number : 146449-90-3
  • Molecular Weight : 208.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-n-pentyloxybenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P280;P271
MSDS https://store.apolloscientific.co.uk/product/4-n-pentyloxybenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart