APO455832805 Apollo Scientific Ltd

4-Amino-3,5-dibromobenzonitrile

  • MDL number MFCD03094688
  • Cas Number : 58633-04-8
  • Molecular Weight : 275.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Cyano-2,6-dibromoaniline
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-amino-35-dibromobenzonitrile

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-amino-35-dibromobenzonitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart