APO455832811 Apollo Scientific Ltd

Pyrimidine-5-boronic acid

  • MDL number MFCD03002366
  • Cas Number : 109299-78-7
  • Molecular Weight : 123.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 110-114
Iupac Name 5-pyrimidinylboronic acid
Inchi Code 1S/C4H5BN2O2/c8-5(9)4-1-6-3-7-2-4/h1-3,8-9H
Inchi Key HZFPPBMKGYINDF-UHFFFAOYSA-N

Description

Synonym 5-Boronopyrimidine, 5-Borono-1,3-diazine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 110-114
Product Link https://store.apolloscientific.co.uk/product/pyrimidine-5-boronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/pyrimidine-5-boronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart