APO455832814 Apollo Scientific Ltd

5-Isobutylcyclohexane-1,3-dione

  • MDL number MFCD00129458
  • Cas Number : 57641-95-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125-129
Synonym 5-(2-Methylprop-1-yl)cyclohexane-1,3-dione, 1,3-Dioxo-5-isobutylcyclohexane
Purity 98
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=14975

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=14975
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart