APO455832819 Apollo Scientific Ltd

4'-Acetyl-[1,1'-biphenyl]-4-carboxylic acid

  • MDL number MFCD00230828
  • Cas Number : 114691-92-8
  • Molecular Weight : 240.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 281-284
Iupac Name 4'-acetyl[1,1'-biphenyl]-4-carboxylic acid
Inchi Code 1S/C15H12O3/c1-10(16)11-2-4-12(5-3-11)13-6-8-14(9-7-13)15(17)18/h2-9H,1H3,(H,17,18)
Inchi Key PDBQNVDCQVGALU-UHFFFAOYSA-N

Description

Synonym 4-(4-Acetylphenyl)benzoic acid, 4'-(4-Carboxyphenyl)acetophenone, 4'-Acetyl-4-carboxybiphenyl
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 281-284
Product Link https://store.apolloscientific.co.uk/product/4-acetyl-11-biphenyl-4-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-acetyl-11-biphenyl-4-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart