APO455832831 Apollo Scientific Ltd

Methyl 1,2-dihydro-2-oxopyridine-4-carboxylate

  • MDL number MFCD09258780
  • Cas Number : 89937-77-9
  • Molecular Weight : 153.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 210-214
Iupac Name methyl 2-oxo-1,2-dihydro-4-pyridinecarboxylate
Inchi Code 1S/C7H7NO3/c1-11-7(10)5-2-3-8-6(9)4-5/h2-4H,1H3,(H,8,9)
Inchi Key DATHOCDTDDUESC-UHFFFAOYSA-N

Description

Synonym Methyl 2-hydroxypyridine-4-carboxylate, 1,2-Dihydro-4-(methoxycarbonyl)-2-oxopyridine
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 210-214
Product Link https://store.apolloscientific.co.uk/product/methyl-12-dihydro-2-oxopyridine-4-carboxylate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-12-dihydro-2-oxopyridine-4-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart