APO455832839 Apollo Scientific Ltd

4-[(2,3-Dihydro-3-oxo-4H-1,4-benzoxazin-4-yl)methyl]benzoic acid

  • MDL number MFCD00269588
  • Cas Number : 857492-98-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(4-Carboxybenzyl)-3,4-dihydro-3-oxo-2H-1,4-benzoxazine, 4-[(2,3-Dihydro-3-oxobenzo[b][1,4]oxazin-4-yl)methyl]benzoic acid
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=13604

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=13604
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart