APO455832849 Apollo Scientific Ltd

2-(Fur-2-yl)benzaldehyde

  • MDL number MFCD03990475
  • Cas Number : 16191-32-5
  • Molecular Weight : 172.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(2-furyl)benzaldehyde
Inchi Code 1S/C11H8O2/c12-8-9-4-1-2-5-10(9)11-6-3-7-13-11/h1-8H
Inchi Key BJGHSPJMFHHOCJ-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fur-2-ylbenzaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-fur-2-ylbenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart